Tisk…

2,7-Dimethyl-3,4-dihydro-2H-1,4-benzoxazine 98%

Dodavatel: Apollo Scientific

OR960211-1G OR960211-5G
APOSOR960211-1GEA 6220 CZK
APOSOR960211-1G APOSOR960211-5G
2,7-Dimethyl-3,4-dihydro-2H-1,4-benzoxazine 98%
2,7-Dimethyl-3,4-dihydro-2H-1,4-benzoxazine
Vzorec: C₁₀H₁₃NO
MW: 163,216 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11603429
CAS číslo: 58960-12-6

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR