Tisk…

2-(Methoxymethyl)pyrrolidine hydrochloride 98%

Dodavatel: Apollo Scientific

OR960192-10G OR960192-1G OR960192-5G
APOSOR960192-10GEA 17990 CZK
APOSOR960192-10G APOSOR960192-1G APOSOR960192-5G
2-(Methoxymethyl)pyrrolidine hydrochloride 98%
2-(Methoxymethyl)pyrrolidine hydrochloride
Vzorec: C₆H₁₃NO·ClH
MW: 151,634 g/mol
Storage Temperature: Ambient
MDL Number: MFCD22056323
CAS číslo: 1376123-33-9

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR