1-(Thiophen-2-ylmethyl)piperidine 98%
Dodavatel: Apollo Scientific
OR960190-10G
OR960190-1G
OR960190-5G
APOSOR960190-10GEA
14290
CZK
APOSOR960190-10G
APOSOR960190-1G
APOSOR960190-5G
1-(Thiophen-2-ylmethyl)piperidine 98%
1-(Thiophen-2-ylmethyl)piperidine
Vzorec:
C₁₀H₁₅NS MW: 181,298 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD18916956 CAS číslo: 91253-06-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...