6-Chloro-4-iodopyridin-3-amine
Dodavatel: Apollo Scientific
OR960084-1G
OR960084-250MG
OR960084-5G
APOSOR960084-1GEA
705
CZK
APOSOR960084-1G
APOSOR960084-250MG
APOSOR960084-5G
6-Chloro-4-iodopyridin-3-amine
6-Chloro-4-iodopyridin-3-amine
Vzorec:
C₅H₄ClIN₂ MW: 254,456 g/mol Storage Temperature: Refrigerator |
MDL Number:
MFCD11520341 CAS číslo: 351227-42-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...