Tisk…

1-(3-Methylbutyl)pyrazol-4-ol 98%

Dodavatel: Apollo Scientific

OR960078-1G OR960078-250MG
APOSOR960078-1GEA 15310 CZK
APOSOR960078-1G APOSOR960078-250MG
1-(3-Methylbutyl)pyrazol-4-ol 98%
1-(3-Methylbutyl)pyrazol-4-ol
Vzorec: C₈H₁₄N₂O
MW: 154.21 g/mol
Storage Temperature: Ambient
MDL Number: MFCD22383778
CAS číslo: 1393442-52-8

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR