Tisk…
2-Ethoxy-5-formylbenzoic acid 96%
Dodavatel: Apollo Scientific
Synonyms: Benzoic acid, 2-ethoxy-5-formyl-
OR960066-1G OR960066-5G
APOSOR960066-1GEA 8270 CZK
APOSOR960066-1G APOSOR960066-5G
2-Ethoxy-5-formylbenzoic acid 96%
2-Ethoxy-5-formylbenzoic acid
Vzorec: C₁₀H₁₀O₄
MW: 194,184 g/mol
Storage Temperature: Ambient
MDL Number: MFCD05625188
CAS číslo: 848606-43-9