Propyl(thiophen-2-ylmethyl)amine 98%
Dodavatel: Apollo Scientific
OR959992-10G
OR959992-1G
OR959992-5G
APOSOR959992-10GEA
14290
CZK
APOSOR959992-10G
APOSOR959992-1G
APOSOR959992-5G
Propyl(thiophen-2-ylmethyl)amine 98%
Propyl(thiophen-2-ylmethyl)amine
Vzorec:
C₈H₁₃NS Storage Temperature: Ambient |
MDL Number:
MFCD04506878 CAS číslo: 751436-48-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...