N-(Thiophen-2-ylmethyl)cyclopentanamine 98%
Dodavatel: Apollo Scientific
OR959965-1G
OR959965-5G
APOSOR959965-1GEA
4770
CZK
APOSOR959965-1G
APOSOR959965-5G
N-(Thiophen-2-ylmethyl)cyclopentanamine 98%
N-(Thiophen-2-ylmethyl)cyclopentanamine
Vzorec:
C₁₀H₁₅NS Storage Temperature: Ambient |
MDL Number:
MFCD03724762 CAS číslo: 58924-50-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...