2-(Diethylamino)benzonitrile 98%
Dodavatel: Apollo Scientific
OR959934-1G
OR959934-5G
APOSOR959934-1GEA
8270
CZK
APOSOR959934-1G
APOSOR959934-5G
2-(Diethylamino)benzonitrile 98%
2-(Diethylamino)benzonitrile
Vzorec:
C₁₁H₁₄N₂ MW: 174,242 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD11155678 CAS číslo: 197507-52-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...