Methyl 1-(cyclopropylmethyl)azetidine-3-carboxylate 95%
Dodavatel: Apollo Scientific
OR959918-1G
OR959918-5G
APOSOR959918-1GEA
6220
CZK
APOSOR959918-1G
APOSOR959918-5G
Methyl 1-(cyclopropylmethyl)azetidine-3-carboxylate 95%
Methyl 1-(cyclopropylmethyl)azetidine-3-carboxylate
Vzorec:
C₉H₁₅NO₂ MW: 169,221 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD20529527 CAS číslo: 1352318-12-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...