Tisk…

3-Hydroxy-2-methoxynaphthalene 98%

Dodavatel: Apollo Scientific

OR959916-1G OR959916-25G OR959916-5G
APOSOR959916-1GEA 3540 CZK
APOSOR959916-1G APOSOR959916-25G APOSOR959916-5G
3-Hydroxy-2-methoxynaphthalene 98%
3-Hydroxy-2-methoxynaphthalene
Vzorec: C₁₂H₁₂O₂
MW: 188,222 g/mol
Storage Temperature: Ambient
MDL Number: MFCD03093901
CAS číslo: 39110-92-4

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR