2-(Methylamino)benzonitrile 98%
Dodavatel: Apollo Scientific
OR959908-1G
OR959908-5G
APOSOR959908-1GEA
2570
CZK
APOSOR959908-1G
APOSOR959908-5G
2-(Methylamino)benzonitrile 98%
2-(Methylamino)benzonitrile
Vzorec:
C₈H₈N₂ MW: 132.163 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD11155692 CAS číslo: 17583-40-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...