Tisk…

2-(Cyclopropylmethyl)pyrazole-3-carbaldehyde 95%

Dodavatel: Apollo Scientific

OR959893-1G OR959893-5G
APOSOR959893-1GEA 9330 CZK
APOSOR959893-1G APOSOR959893-5G
2-(Cyclopropylmethyl)pyrazole-3-carbaldehyde 95%
2-(Cyclopropylmethyl)pyrazole-3-carbaldehyde
Vzorec: C₈H₁₀N₂O
Storage Temperature: Ambient
MDL Number: MFCD18251009
CAS číslo: 1236365-97-1

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR