N-Methoxy-N,2-dimethylpropanamide 98%
Dodavatel: Apollo Scientific
Synonyms:
N-Methoxy-N-methylisobutyramide
OR959864-25G
APOSOR959864-25GEA
3970
CZK
APOSOR959864-25G
N-Methoxy-N,2-dimethylpropanamide 98%
N-Methoxy-N,2-dimethylpropanamide
Vzorec:
C₆H₁₃NO₂ MW: 131,173 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD11847606 CAS číslo: 113778-69-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...