Tisk…

4-cyanoquinoline 97%

Dodavatel: Apollo Scientific

Synonyms: Quinoline-4-carbonitrile

OR959787-1G
APOSOR959787-1GEA 1490 CZK
APOSOR959787-1G
4-cyanoquinoline 97%
4-cyanoquinoline
Vzorec: C₁₀H₆N₂
MW: 154,168 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00972111
CAS číslo: 2973-27-5

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR