b-Amino-3-chlorobenzenepropanol 97%
Dodavatel: Apollo Scientific
OR959761-1G
OR959761-250MG
OR959761-500MG
APOSOR959761-1GEA
22320
CZK
APOSOR959761-1G
APOSOR959761-250MG
APOSOR959761-500MG
b-Amino-3-chlorobenzenepropanol 97%
b-Amino-3-chlorobenzenepropanol
Vzorec:
C₉H₁₂ClNO Storage Temperature: Ambient |
MDL Number:
MFCD10034619 CAS číslo: 37844-06-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...