Tisk…

N,3-Dimethylpyridine-2-carboxamide 98%

Dodavatel: Apollo Scientific

OR959755-1G OR959755-5G
APOSOR959755-1GEA 3740 CZK
APOSOR959755-1G APOSOR959755-5G
N,3-Dimethylpyridine-2-carboxamide 98%
N,3-Dimethylpyridine-2-carboxamide
Vzorec: C₈H₁₀N₂O
Storage Temperature: Ambient
MDL Number: MFCD19671808
CAS číslo: 1037045-67-2

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR