Tisk…

Ethyl 2-(N-hydroxyimino)-2-(pyridin-2-yl)acetate 98%

Dodavatel: Apollo Scientific

OR959749-1G OR959749-250MG
APOSOR959749-1GEA 8270 CZK
APOSOR959749-1G APOSOR959749-250MG
Ethyl 2-(N-hydroxyimino)-2-(pyridin-2-yl)acetate 98%
Ethyl 2-(N-hydroxyimino)-2-(pyridin-2-yl)acetate
Vzorec: C₉H₁₀N₂O₃
MW: 194,187 g/mol
Storage Temperature: Ambient
MDL Number: MFCD03939045
CAS číslo: 71721-67-0

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR