Tisk…

7-Nitro-3,4-dihydro-2H-isoquinolin-1-one 95%

Dodavatel: Apollo Scientific

Synonyms: 7-Nitro-3,4-dihydroisoquinolin-1(2H)-one

OR959733-100MG OR959733-1G OR959733-250MG
APOSOR959733-100MGEA 1740 CZK
APOSOR959733-100MG APOSOR959733-1G APOSOR959733-250MG
7-Nitro-3,4-dihydro-2H-isoquinolin-1-one 95%
7-Nitro-3,4-dihydro-2H-isoquinolin-1-one
Vzorec: C₉H₈N₂O₃
MW: 192,171 g/mol
Storage Temperature: Ambient
MDL Number: MFCD08437646
CAS číslo: 22245-96-1

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR