Tisk…

5-Chloroquinazolin-4-amine 98%

Dodavatel: Apollo Scientific

OR959731-1G OR959731-250MG
APOSOR959731-1GEA 15310 CZK
APOSOR959731-1G APOSOR959731-250MG
5-Chloroquinazolin-4-amine 98%
5-Chloroquinazolin-4-amine
Vzorec: C₈H₆ClN₃
MW: 179,606 g/mol
Storage Temperature: Ambient
MDL Number: MFCD02323641
CAS číslo: 19808-34-5

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR