2-Chloro-5-methylpyridin-4-ol 98%
Dodavatel: Apollo Scientific
OR959693-1G
OR959693-250MG
APOSOR959693-1GEA
17990
CZK
APOSOR959693-1G
APOSOR959693-250MG
2-Chloro-5-methylpyridin-4-ol 98%
2-Chloro-5-methylpyridin-4-ol
Vzorec:
C₆H₆ClNO MW: 143,571 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD16610469 CAS číslo: 1227580-80-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...