Tisk…

Methyl 5-methoxypyridine-2-carboximidate 98%

Dodavatel: Apollo Scientific

OR959632-1G OR959632-5G
APOSOR959632-1GEA 7670 CZK
APOSOR959632-1G APOSOR959632-5G
Methyl 5-methoxypyridine-2-carboximidate 98%
Methyl 5-methoxypyridine-2-carboximidate
Vzorec: C₈H₁₀N₂O₂
Storage Temperature: Ambient
MDL Number: MFCD25953920
CAS číslo: 1820588-49-5

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR