Tisk…

1-(5-Methoxypyridin-2-yl)ethanone 96%

Dodavatel: Apollo Scientific

OR959601-1G OR959601-250MG
APOSOR959601-1GEA 15710 CZK
APOSOR959601-1G APOSOR959601-250MG
1-(5-Methoxypyridin-2-yl)ethanone 96%
1-(5-Methoxypyridin-2-yl)ethanone
Vzorec: C₈H₉NO₂
Storage Temperature: Ambient
MDL Number: MFCD10697709
CAS číslo: 325796-84-7

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR