3-Chloro-6-methoxy-2-methylpyridine 98%
Dodavatel: Apollo Scientific
OR959592-1G
OR959592-5G
APOSOR959592-1GEA
6220
CZK
APOSOR959592-1G
APOSOR959592-5G
3-Chloro-6-methoxy-2-methylpyridine 98%
3-Chloro-6-methoxy-2-methylpyridine
Vzorec:
C₇H₈ClNO Storage Temperature: Ambient |
MDL Number:
MFCD16610601 CAS číslo: 1227593-97-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...