Tisk…

4-Methoxy-N-methylpyrimidin-2-amine 97%

Dodavatel: Apollo Scientific

OR959581-1G OR959581-250MG
APOSOR959581-1GEA 22290 CZK
APOSOR959581-1G APOSOR959581-250MG
4-Methoxy-N-methylpyrimidin-2-amine 97%
4-Methoxy-N-methylpyrimidin-2-amine
Vzorec: C₆H₉N₃O
MW: 139,155 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09054909
CAS číslo: 66131-71-3

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR