4-Amino-2-mercaptopyrimidine-5-carbonitrile
Dodavatel: Apollo Scientific
|
Varování
|
OR959533-1G
OR959533-5G
APOSOR959533-1GEA
3310
CZK
APOSOR959533-1G
APOSOR959533-5G
4-Amino-2-mercaptopyrimidine-5-carbonitrile
4-Amino-2-mercaptopyrimidine-5-carbonitrile
Vzorec:
C₅H₄N₄S MW: 152,177 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00085719 CAS číslo: 16462-26-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...