Tisk…

N,N-Dimethyl-5-nitropyridin-2-amine 97%

Dodavatel: Apollo Scientific

OR959529-1G OR959529-25G OR959529-5G
APOSOR959529-1GEA 1540 CZK
APOSOR959529-1G APOSOR959529-25G APOSOR959529-5G
N,N-Dimethyl-5-nitropyridin-2-amine 97%
N,N-Dimethyl-5-nitropyridin-2-amine
Vzorec: C₇H₉N₃O₂
MW: 167,165 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00023450
CAS číslo: 2554-75-8

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR