Tisk…

5-Chloro-1-methylpyridin-2-one 98%

Dodavatel: Apollo Scientific

OR959475-1G OR959475-5G
APOSOR959475-1GEA 4900 CZK
APOSOR959475-1G APOSOR959475-5G
5-Chloro-1-methylpyridin-2-one 98%
5-Chloro-1-methylpyridin-2-one
Vzorec: C₆H₆ClNO
MW: 143,571 g/mol
Storage Temperature: Ambient
MDL Number: MFCD22375100
CAS číslo: 4214-78-2

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR