Tisk…

3-Bromo-4-ethoxypyridine 95%

Dodavatel: Apollo Scientific

OR959462-1G OR959462-250MG
APOSOR959462-1GEA 13460 CZK
APOSOR959462-1G APOSOR959462-250MG
3-Bromo-4-ethoxypyridine 95%
3-Bromo-4-ethoxypyridine
Vzorec: C₇H₈BrNO
MW: 202,049 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00234192
CAS číslo: 3522-97-2

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR