Tisk…

5-Bromo-2-methylnicotinic acid 95%

Dodavatel: Apollo Scientific
Varování

OR959451-5G
APOSOR959451-5GEA 4860 CZK
APOSOR959451-5G
5-Bromo-2-methylnicotinic acid 95%
5-Bromo-2-methylnicotinic acid
Vzorec: C₇H₆BrNO₂
MW: 216,032 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11557230
CAS číslo: 351003-02-6

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR