Tisk…

N-Cyclopropyl 4-chloropicolinamide 98%

Dodavatel: Apollo Scientific

OR959440-1G OR959440-5G
APOSOR959440-1GEA 6220 CZK
APOSOR959440-1G APOSOR959440-5G
N-Cyclopropyl 4-chloropicolinamide 98%
N-Cyclopropyl 4-chloropicolinamide
Vzorec: C₉H₉ClN₂O
Storage Temperature: Ambient
MDL Number: MFCD10663418
CAS číslo: 1090815-16-9

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR