Tisk…

3-Oxabicyclo[3.1.0]hexan-6-amine hydrochloride 97%

Dodavatel: Apollo Scientific
Varování

OR959373-100MG OR959373-250MG
APOSOR959373-100MGEA 3740 CZK
APOSOR959373-100MG APOSOR959373-250MG
3-Oxabicyclo[3.1.0]hexan-6-amine hydrochloride 97%
3-Oxabicyclo[3.1.0]hexan-6-amine hydrochloride
Vzorec: C₅H₉NO·ClH
MW: 135,592 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD13193583
CAS číslo: 693248-55-4

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR