Tisk…

2-Cyano-2-methylacetic acid 95%

Dodavatel: Apollo Scientific

OR959297-1G OR959297-250MG
APOSOR959297-1GEA 10150 CZK
APOSOR959297-1G APOSOR959297-250MG
2-Cyano-2-methylacetic acid 95%
2-Cyano-2-methylacetic acid
Vzorec: C₄H₅NO₂
Bod varu: 115…117 °C (2 mm Hg)
Bod tání: 35…36 °C
Hustota: 1,14 g/cm³ (25 °C)
Storage Temperature: Refrigerator
MDL Number: MFCD14544816
CAS číslo: 632-07-5
UN: 3265
ADR: 8,II

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR