Tisk…

(R)-3-(1-(Dimethylamino)ethyl)phenol 95%

Dodavatel: Apollo Scientific

OR959139-5G
APOSOR959139-5GEA 2610 CZK
APOSOR959139-5G
(R)-3-(1-(Dimethylamino)ethyl)phenol 95%
(R)-3-(1-(Dimethylamino)ethyl)phenol
Vzorec: C₁₀H₁₅NO
MW: 165.232 g/mol
Storage Temperature: Ambient
MDL Number: MFCD06656490
CAS číslo: 851086-95-8

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR