Tisk…
But-2-ene-1,4-diamine, N-BOC protected 95%
Dodavatel: Apollo Scientific
Nebezpečí
Synonyms: (E)-tert-Butyl (4-aminobut-2-en-1-yl)carbamate
OR958835-1G OR958835-250MG
APOSOR958835-1GEA 4700 CZK
APOSOR958835-1G APOSOR958835-250MG
But-2-ene-1,4-diamine, N-BOC protected 95%
But-2-ene-1,4-diamine, N-BOC protected
Vzorec: C₉H₁₈N₂O₂
MW: 186,251 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD09752966
CAS číslo: 146394-99-2
UN: 2735
ADR: 8,III