Tisk…

Ethyl 2-(6-bromopyridin-2-yl)acetate

Dodavatel: Apollo Scientific

OR958798-1G OR958798-5G OR958798-25G
APOSOR958798-1GEA 4240 CZK
APOSOR958798-1G APOSOR958798-5G APOSOR958798-25G
Ethyl 2-(6-bromopyridin-2-yl)acetate
Ethyl 2-(6-bromopyridin-2-yl)acetate
Vzorec: C₉H₁₀BrNO₂
MW: 244,085 g/mol
Storage Temperature: Ambient
MDL Number: MFCD10566284
CAS číslo: 955369-63-8

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR