Tisk…

(S)-b-Amino-3-methoxy-benzeneethanol 97%

Dodavatel: Apollo Scientific

Synonyms: (S)-2-Amino-2-(3-methoxyphenyl)ethanol

OR958678-1G OR958678-250MG OR958678-500MG
APOSOR958678-1GEA 24800 CZK
APOSOR958678-1G APOSOR958678-250MG APOSOR958678-500MG
(S)-b-Amino-3-methoxy-benzeneethanol 97%
(S)-b-Amino-3-methoxy-benzeneethanol
Vzorec: C₉H₁₃NO₂
MW: 167.205 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09253686
CAS číslo: 1213016-49-9

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR