2-Methoxy-6-methylpyridin-4-amine 95%
Dodavatel: Apollo Scientific
OR958656-1G
APOSOR958656-1GEA
3440
CZK
APOSOR958656-1G
2-Methoxy-6-methylpyridin-4-amine 95%
2-Methoxy-6-methylpyridin-4-amine
Vzorec:
C₇H₁₀N₂O MW: 138,167 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD15529703 CAS číslo: 89943-09-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...