Tisk…

2-(2-Phenylethynyl)aniline 95%

Dodavatel: Apollo Scientific

Synonyms: 2-Phenylethynyl-phenylamine

OR958600-1G OR958600-250MG
APOSOR958600-1GEA 2570 CZK
APOSOR958600-1G APOSOR958600-250MG
2-(2-Phenylethynyl)aniline 95%
2-(2-Phenylethynyl)aniline
Vzorec: C₁₄H₁₁N
MW: 193,244 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00168849
CAS číslo: 13141-38-3

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR