2-(Phenylethynyl)benzaldehyde 98%
Dodavatel: Apollo Scientific
OR958562-1G
OR958562-250MG
OR958562-5G
APOSOR958562-1GEA
3020
CZK
APOSOR958562-1G
APOSOR958562-250MG
APOSOR958562-5G
2-(Phenylethynyl)benzaldehyde 98%
2-(Phenylethynyl)benzaldehyde
Vzorec:
C₁₅H₁₀O MW: 206,239 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD09030317 CAS číslo: 59046-72-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...