Tisk…

8-Chloro-4-methylquinolin-2-ol 95%

Dodavatel: Apollo Scientific

OR958392-100MG OR958392-1G OR958392-250MG
APOSOR958392-100MGEA 1660 CZK
APOSOR958392-100MG APOSOR958392-1G APOSOR958392-250MG
8-Chloro-4-methylquinolin-2-ol 95%
8-Chloro-4-methylquinolin-2-ol
Vzorec: C₁₀H₈ClNO
Storage Temperature: Ambient
MDL Number: MFCD11868958
CAS číslo: 54904-38-0

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR