Tisk…

Ethyl 2-(2-hydroxyphenoxy)acetate 95%

Dodavatel: Apollo Scientific

OR958223-1G OR958223-5G
APOSOR958223-1GEA 2030 CZK
APOSOR958223-1G APOSOR958223-5G
Ethyl 2-(2-hydroxyphenoxy)acetate 95%
Ethyl 2-(2-hydroxyphenoxy)acetate
Vzorec: C₁₀H₁₂O₄
MW: 196.2 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00547114
CAS číslo: 99186-63-7

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR