Tisk…

3-Amino-2-bromobenzonitrile 95%

Dodavatel: Apollo Scientific

OR958189-1G OR958189-250MG
APOSOR958189-1GEA 1700 CZK
APOSOR958189-1G APOSOR958189-250MG
3-Amino-2-bromobenzonitrile 95%
3-Amino-2-bromobenzonitrile
Vzorec: C₇H₅BrN₂
MW: 197,032 g/mol
Storage Temperature: Ambient
MDL Number: MFCD13185832
CAS číslo: 1166988-09-5

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR