(4S)-6-Chlorochroman-4-amine hydrochloride
Dodavatel: Apollo Scientific
|
Varování
|
Synonyms:
(S)-6-Chlorochroman-4-amine hydrochloride, (4S)-6-chloro-3,4-dihydro-2H-chromen-4-amine, hydrochloride
OR958124-250MG
OR958124-1G
APOSOR958124-250MGEA
8300
CZK
APOSOR958124-250MG
APOSOR958124-1G
(4S)-6-Chlorochroman-4-amine hydrochloride
(4S)-6-Chlorochroman-4-amine hydrochloride
Vzorec:
C₉H₁₀ClNO·ClH MW: 220,096 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD24435622 CAS číslo: 1956436-48-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...