Tisk…

2,4-Bis(methylthio)pyrimidine 95%

Dodavatel: Apollo Scientific

OR958110-1G OR958110-250MG
APOSOR958110-1GEA 2200 CZK
APOSOR958110-1G APOSOR958110-250MG
2,4-Bis(methylthio)pyrimidine 95%
2,4-Bis(methylthio)pyrimidine
Vzorec: C₆H₈N₂S₂
MW: 172,271 g/mol
Storage Temperature: Ambient
MDL Number: MFCD18974850
CAS číslo: 5909-26-2

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR