Tisk…

3,6-Dimethylpicolinic acid 95%

Dodavatel: Apollo Scientific

OR958005-1G OR958005-250MG
APOSOR958005-1GEA 13860 CZK
APOSOR958005-1G APOSOR958005-250MG
3,6-Dimethylpicolinic acid 95%
3,6-Dimethylpicolinic acid
Vzorec: C₈H₉NO₂
Storage Temperature: Ambient
MDL Number: MFCD12401109
CAS číslo: 83282-46-6

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR