Tisk…

4-Butyl-2-nitroaniline 95%

Dodavatel: Apollo Scientific

OR957993-5G
APOSOR957993-5GEA 3440 CZK
APOSOR957993-5G
4-Butyl-2-nitroaniline 95%
4-Butyl-2-nitroaniline
Vzorec: C₁₀H₁₄N₂O₂
MW: 194,23 g/mol
Storage Temperature: Ambient
MDL Number: MFCD23725964
CAS číslo: 3663-22-7

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR