2-Chloro-5-isopropoxypyridine 95%
Dodavatel: Apollo Scientific
OR957933-1G
OR957933-250MG
APOSOR957933-1GEA
4270
CZK
APOSOR957933-1G
APOSOR957933-250MG
2-Chloro-5-isopropoxypyridine 95%
2-Chloro-5-isopropoxypyridine
Vzorec:
C₈H₁₀ClNO MW: 171.624 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD19689743 CAS číslo: 1204483-99-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...