Tisk…

4-Chloro-2-(chloromethyl)pyridine hydrochloride 95%

Dodavatel: Apollo Scientific

OR957923-1G
APOSOR957923-1GEA 4470 CZK
APOSOR957923-1G
4-Chloro-2-(chloromethyl)pyridine hydrochloride 95%
4-Chloro-2-(chloromethyl)pyridine hydrochloride
Vzorec: C₆H₅Cl₂N·ClH
MW: 198,478 g/mol
Storage Temperature: Ambient
MDL Number: MFCD08234868
CAS číslo: 119396-04-2

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR