5-Methyl-6-nitro-1,3-benzodioxole 95%
Dodavatel: Apollo Scientific
OR957738-1G
OR957738-250MG
APOSOR957738-1GEA
7670
CZK
APOSOR957738-1G
APOSOR957738-250MG
5-Methyl-6-nitro-1,3-benzodioxole 95%
5-Methyl-6-nitro-1,3-benzodioxole
Vzorec:
C₈H₇NO₄ Storage Temperature: Ambient |
MDL Number:
MFCD00229114 CAS číslo: 32996-27-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...